Gene Transcripts

Gene Symbol
Cacna2d4
Entrez Gene ID
319734
Gene Name
calcium channel, voltage-dependent, alpha 2/delta subunit 4
Gene Aliases
5730412N02Rik, BE686333
Chromosome Location
Chr. 6: 119236474 - 119353392 on Build GRCm38
UniGene ID
Mm.440773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033382.2 NP_001028554.2 36, 37 3332
GenBank mRNA AK137847.1 - 10, 11 638
BC141091.1 - 35, 36 3188 9055703

View Full Product Details