Gene Transcripts

Gene Symbol
Sult2a6
Entrez Gene ID
629219
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 6
Gene Aliases
EG629219, Gm6957
Chromosome Location
Chr. 7: 14222403 - 14254870 on Build GRCm38
UniGene ID
Mm.439627
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081325.2 NP_001074794.1 1 147
GenBank mRNA BC147210.1 - 1 147
BC172008.1 - 1 147

View Full Product Details