Gene Transcripts

Gene Symbol
Kbtbd11
Entrez Gene ID
74901
Gene Name
kelch repeat and BTB (POZ) domain containing 11
Gene Aliases
2900016B01Rik, 4930465M17Rik, mKIAA0711
Chromosome Location
Chr. 8: 15011011 - 15033333 on Build GRCm38
UniGene ID
Mm.46675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029116.2 NP_083392.1 2 1758
GenBank mRNA AK050286.1 - 1 613
AK080001.1 - 1 777
AK129197.1 - 1 721
BC080858.1 - 1 729 6852896

View Full Product Details