Product Details
- SNP ID
-
rs13483330
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:43764720 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTCATCGGCTATAAAGACTGTTG[C/T]AAGTAAGCTTATAGTAGGATGAGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Scgb3a2
PubMed Links
Gene Details
- Gene
- Scgb3a2
- Gene Name
- secretoglobin, family 3A, member 2
There are no transcripts associated with this gene.
View Full Product Details