Product Details
- SNP ID
-
rs13481788
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:43632644 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACATGACAAATTCAATGTTTTTTT[C/T]CCCTCTGTCTCTACAGTTGAAGATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rnf182
PubMed Links
Gene Details
- Gene
- Rnf182
- Gene Name
- ring finger protein 182
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183204.4 |
|
Intron |
|
|
NP_899027.1 |
View Full Product Details