Product Details
- SNP ID
-
rs13482468
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:20818544 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGTCCTGGGGTAGAATTCAGAT[G/T]GAATAATTTTAGCCAAGAAGAACTT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Cdh12
PubMed Links
Gene Details
- Gene
- Cdh12
- Gene Name
- cadherin 12
View Full Product Details