Product Details

SNP ID
rs6323325
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:82406934 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAATGACAGATGTGTAAAGCCATGA[C/T]GAGTATTCTACCCATTCTGACTTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Nfib PubMed Links

Gene Details

Gene
Nfib
Gene Name
nuclear factor I/B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113209.2 Intron NP_001106680.1
NM_001113210.2 Intron NP_001106681.1
NM_001286127.1 Intron NP_001273056.1
NM_001286131.1 Intron NP_001273060.1
NM_008687.6 Intron NP_032713.3

View Full Product Details