Product Details

SNP ID
rs3700249
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:66161850 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTCTTTCAGAGTTCCTTTATGACA[G/A]TCACTTCTAAACGCCTGCCACAAAA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Chrna2 PubMed Links

Gene Details

Gene
Chrna2
Gene Name
cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal)
There are no transcripts associated with this gene.

Gene
Ptk2b
Gene Name
PTK2 protein tyrosine kinase 2 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162365.1 Intron NP_001155837.1
NM_001162366.1 Intron NP_001155838.1
NM_172498.3 Intron NP_766086.2

View Full Product Details