Product Details
- SNP ID
-
rs13459055
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:179043861 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGATGTTGATGTCGGGAAGCCGGT[C/T]GGAATTGCTGTTTATGATCGCCTGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Smyd3
PubMed Links
Gene Details
- Gene
- Smyd3
- Gene Name
- SET and MYND domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_027188.3 |
1004 |
Missense Mutation |
|
|
NP_081464.1 |
View Full Product Details