Product Details
- SNP ID
-
rs13477828
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:88196852 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACGTCATTGGAGAAGAGTCTGCC[A/G]CTAGTGACTGGCTGGCTACAGTGGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Focad
PubMed Links
Gene Details
- Gene
- Focad
- Gene Name
- focadhesin
View Full Product Details