Product Details
- SNP ID
-
rs13483108
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75512026 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAATAAGCCTGTGGTACCTCTGGA[A/G]TGGGCTTCAGAAGTGGTACCAAAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rasgrp3
PubMed Links
Gene Details
- Gene
- Rasgrp3
- Gene Name
- RAS, guanyl releasing protein 3
View Full Product Details