Product Details

SNP ID
rs13475873
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:53213120 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCTAAAGAAACTACATAGTGA[A/C]AATGTATCACAAATCAAGTACAATT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Pms1 PubMed Links

Gene Details

Gene
Pms1
Gene Name
postmeiotic segregation increased 1 (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153556.1 Intron NP_705784.1

View Full Product Details