Product Details

SNP ID
rs13481826
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:57558363 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTTTCTCATCTATAAGTGAAG[A/G]AAACAGAAACTACATCTGGTGGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Spock1 PubMed Links

Gene Details

Gene
Spock1
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166463.1 Intron NP_001159935.1
NM_001166464.1 Intron NP_001159936.1
NM_001166465.1 Intron NP_001159937.1
NM_001166466.1 Intron NP_001159938.1
NM_009262.3 Intron NP_033288.2

View Full Product Details