Product Details
- SNP ID
-
rs13481890
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:77046571 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATCTCCACATGTCCGTTTGACAG[C/T]GCATCATGCAAAGGCGTCACCCCGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ankrd32
PubMed Links
Gene Details
- Gene
- Ankrd32
- Gene Name
- ankyrin repeat domain 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_134071.3 |
2750 |
Silent Mutation |
|
|
NP_598832.3 |
View Full Product Details