Product Details
- SNP ID
-
rs13480677
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:84580684 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAAAGCTGGTGAATCTTTATAAT[C/T]CTCAGCCAGTACAGGGTCATTTATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tcp11l2
PubMed Links
Gene Details
- Gene
- Tcp11l2
- Gene Name
- t-complex 11 (mouse) like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_146008.2 |
|
Intron |
|
|
NP_666120.1 |
View Full Product Details