Product Details

SNP ID
rs13482488
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:26376747 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGAGGTTTTCTGCCTTTTGCT[A/C]TCATCCTGTGTTATGAAGGTCACTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
March11 PubMed Links

Gene Details

Gene
March11
Gene Name
membrane-associated ring finger (C3HC4) 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177597.5 Intron NP_808265.2

View Full Product Details