Product Details

SNP ID
rs29405909
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:55483148 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCTTCACTATGCACCACTGCCGCC[A/G]CCTCCCTCCCATATTTTAAACAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G3bp1 PubMed Links

Gene Details

Gene
G3bp1
Gene Name
GTPase activating protein (SH3 domain) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013716.2 Intron NP_038744.1
Gene
Gm12238
Gene Name
predicted gene 12238
There are no transcripts associated with this gene.

View Full Product Details