Product Details
- SNP ID
-
rs3662712
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:16687131 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGACCTTTTATCATCTAAAATACA[G/A]TTTTTTACTGAGAAAGAAAAGACTG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Vps13a
PubMed Links
Gene Details
- Gene
- Vps13a
- Gene Name
- vacuolar protein sorting 13A (yeast)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173028.4 |
4511 |
UTR 3 |
|
|
NP_766616.2 |
View Full Product Details