Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HJV
Entrez Gene ID
102138915
Gene Name
hemojuvelin BMP co-receptor
Gene Aliases
HFE2
Chromosome Location
Chr.1: 104304859 - 104309315 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005542086.2 XP_005542143.1 1 - 2 340 73

View Full Product Details