Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLRC1
Entrez Gene ID
102121106
Gene Name
killer cell lectin like receptor C1
Gene Aliases
nkg2A
Chromosome Location
Chr.11: 10790440 - 10804294 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284177.1 NP_001271106.1 3 - 4 354 92
XM_015430169.1 XP_015285655.1 4 - 5 1513 92
XM_015430170.1 XP_015285656.1 5 - 6 1956 92
XM_015430171.1 XP_015285657.1 4 - 5 1513 92
XM_015430172.1 XP_015285658.1 5 - 6 1956 92
GenBank mRNA AJ585528.1 - 3 - 4 354 92

View Full Product Details