Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFI27L2
Entrez Gene ID
102131700
Gene Name
interferon alpha inducible protein 27 like 2
Gene Aliases
-
Chromosome Location
Chr.7: 159178597 - 159180505 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005562096.2 XP_005562153.1 3 - 4 299 124

View Full Product Details