Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRG1
Entrez Gene ID
102130067
Gene Name
killer cell lectin like receptor G1
Gene Aliases
-
Chromosome Location
Chr.11: 9273047 - 9294005 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570060.2 XP_005570117.1 1 - 2 228 75

View Full Product Details