Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITLN1
Entrez Gene ID
102132072
Gene Name
intelectin 1
Gene Aliases
-
Chromosome Location
Chr.1: 90749527 - 90758718 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541233.2 XP_005541290.1 1 - 2 192 60

View Full Product Details