Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102128656
Entrez Gene ID
102128656
Gene Name
centrosomal protein C10orf90
Gene Aliases
C9H10orf90
Chromosome Location
Chr.9: 125757631 - 125846399 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566724.2 XP_005566781.1 4 - 5 1974 121
XM_015456855.1 XP_015312341.1 5 - 6 2260 121

View Full Product Details