Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARX
Entrez Gene ID
102136734
Gene Name
aristaless related homeobox
Gene Aliases
-
Chromosome Location
Chr.X: 23017863 - 23030130 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015443299.1 XP_015298785.1 5 - 6 1475 122

View Full Product Details