Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNJ11
Entrez Gene ID
102120776
Gene Name
potassium inwardly rectifying channel subfamily J member 11
Gene Aliases
-
Chromosome Location
Chr.14: 50341017 - 50344895 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005578496.1 XP_005578553.1 1 - 1 3214 84

View Full Product Details