Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC42EP2
Entrez Gene ID
102143427
Gene Name
CDC42 effector protein 2
Gene Aliases
-
Chromosome Location
Chr.14: 9201990 - 9210425 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577296.2 XP_005577353.1 1 - 2 137 98

View Full Product Details