Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NME6
Entrez Gene ID
102139844
Gene Name
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases
-
Chromosome Location
Chr.2: 103384718 - 103392199 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284952.1 NP_001271881.1 6 - 7 633 72
GenBank mRNA AB172966.1 - 6 - 7 633 72

View Full Product Details