Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OBP2B
Entrez Gene ID
102134500
Gene Name
odorant binding protein 2B
Gene Aliases
-
Chromosome Location
Chr.15: 5555912 - 5565532 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015436458.1 XP_015291944.1 4 - 5 431 95

View Full Product Details