Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL2L10
Entrez Gene ID
102126425
Gene Name
BCL2 like 10
Gene Aliases
-
Chromosome Location
Chr.7: 29107862 - 29111302 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005559578.2 XP_005559635.1 1 - 2 473 99

View Full Product Details