Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PATE4
Entrez Gene ID
102142771
Gene Name
prostate and testis expressed 4
Gene Aliases
-
Chromosome Location
Chr.14: 121667765 - 121675255 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580094.2 XP_005580151.1 2 - 3 253 131

View Full Product Details