Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTBD6
Entrez Gene ID
102120929
Gene Name
BTB domain containing 6
Gene Aliases
-
Chromosome Location
Chr.7: 170614289 - 170616256 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005562383.2 XP_005562440.1 2 - 3 253 107

View Full Product Details