Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH4A
Entrez Gene ID
102119712
Gene Name
radial spoke head component 4A
Gene Aliases
-
Chromosome Location
Chr.4: 20640972 - 20658032 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551656.2 XP_005551713.1 2 - 3 1424 100
XM_005551657.2 XP_005551714.1 2 - 3 1424 100

View Full Product Details