Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLCO1B7
Entrez Gene ID
102136968
Gene Name
solute carrier organic anion transporter family member 1B7 (putative)
Gene Aliases
-
Chromosome Location
Chr.11: 21721179 - 21806370 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015430187.1 XP_015285673.1 5 - 6 470 92

View Full Product Details