Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYF6
Entrez Gene ID
102127495
Gene Name
myogenic factor 6
Gene Aliases
-
Chromosome Location
Chr.11: 79874469 - 79876543 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571633.2 XP_005571690.1 1 - 2 740 57

View Full Product Details