The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044744.1 | NP_001038209.2 | 2 - 3 | 309 | 68 |
NM_008097.2 | NP_032123.3 | 1 - 2 | 366 | 68 | |
GenBank mRNA | AK165406.1 | - | 1 - 2 | 192 | 68 |
AU080952.1 | - | 2 - 3 | 166 | 68 | |
BC061158.1 | - | 1 - 2 | 196 | 68 | |
BG807173.1 | - | 1 - 2 | 362 | 68 | |
BY301880.1 | - | 2 - 3 | 310 | 68 | |
U18992.1 | - | 1 - 2 | 147 | 68 |