The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271628.1 | NP_001258557.1 | 3 - 3 | 732 | 84 |
NM_008121.3 | NP_032147.1 | 2 - 2 | 675 | 84 | |
GenBank mRNA | AK038780.1 | - | - | 1404 | 84 |
AK137069.1 | - | 2 - 2 | 637 | 84 | |
BC053054.1 | - | 2 - 2 | 661 | 84 |