The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008646.3 | NP_032672.2 | 16 - 17 | 2048 | 117 |
GenBank mRNA | M65238.1 | - | 16 - 17 | 2051 | 117 |