The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190187.1 | NP_001177116.1 | 5 - 6 | 1443 | 94 |
NM_001190188.1 | NP_001177117.1 | 5 - 6 | 1446 | 94 | |
NM_008734.3 | NP_032760.1 | 5 - 6 | 1446 | 94 | |
GenBank mRNA | AF010130.1 | - | 5 - 6 | 1446 | 94 |
BC137838.1 | - | 5 - 6 | 1177 | 94 | |
BC144964.1 | - | 5 - 6 | 1183 | 94 |