The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159724.1 | NP_001153196.1 | 1 - 2 | 362 | 55 |
NM_008990.3 | NP_033016.3 | 1 - 2 | 362 | 55 | |
GenBank mRNA | AK075815.1 | - | 1 - 2 | 267 | 55 |
AK157513.1 | - | 1 - 2 | 267 | 55 | |
BC009088.1 | - | 1 - 2 | 255 | 55 | |
BC046770.1 | - | 1 - 2 | 258 | 55 | |
BC059941.1 | - | 1 - 2 | 314 | 55 | |
D26107.1 | - | 1 - 2 | 91 | 55 | |
M80206.1 | - | 1 - 2 | 91 | 55 |