The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009511.2 | NP_033537.1 | 1 - 2 | 168 | 63 |
GenBank mRNA | AK038743.1 | - | 1 - 2 | 169 | 63 |
AK142357.1 | - | 1 - 2 | 134 | 63 | |
BC138572.1 | - | 1 - 2 | 66 | 63 | |
D28132.1 | - | 1 - 2 | 100 | 63 |