Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B2m
Entrez Gene ID
12010
Gene Name
beta-2 microglobulin
Gene Aliases
Ly-m11, beta2-m, beta2m
Chromosome Location
Chr.2: 122147687 - 122153082 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009735.3 NP_033865.2 3 - 4 426 125
GenBank mRNA AK019389.1 - 3 - 4 429 125
AK020632.1 - 3 - 4 428 125
AK028378.1 - 3 - 4 428 125
AK153255.1 - 3 - 4 428 125
AK166067.1 - 3 - 4 429 125
BC085164.1 - 3 - 4 381 125
X01838.1 - 3 - 4 427 125

View Full Product Details