The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009793.3 | NP_033923.2 | 1 - 2 | 502 | 68 |
GenBank mRNA | AK030219.1 | - | 1 - 2 | 386 | 68 |
AK043114.1 | - | 1 - 2 | 212 | 68 | |
BC070420.1 | - | 1 - 2 | 383 | 68 | |
X58995.1 | - | 1 - 2 | 192 | 68 |