Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dab1
Entrez Gene ID
13131
Gene Name
disabled 1
Gene Aliases
AI956902, C630028C02Rik, scm, scr, scrambler, yot
Chromosome Location
Chr.4: 103619546 - 104744844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010014.3 NP_034144.1 3 - 4 519 118
NM_177259.4 NP_796233.2 3 - 4 519 118
NR_104385.1 3 - 4 519 118
GenBank mRNA AK005640.1 - 2 - 3 330 118
BC051993.1 - 5 - 6 652 118
BC138538.1 - 2 - 3 240 118
BC138539.1 - 2 - 3 240 118
BY731247.1 - 3 - 4 486 118
Y08379.1 - 3 - 4 467 118
Y08380.1 - 3 - 4 467 118
Y08381.1 - 3 - 4 467 118

View Full Product Details