Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itih2
Entrez Gene ID
16425
Gene Name
inter-alpha trypsin inhibitor, heavy chain 2
Gene Aliases
AI747202, Intin2, Itih-2
Chromosome Location
Chr.2: 10094591 - 10130683 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010582.3 NP_034712.2 1 - 2 197 69
GenBank mRNA AK133782.1 - 1 - 2 96 69
AK149533.1 - 2 - 3 242 69
BC034341.1 - 1 - 2 171 69
BY776793.1 - 1 - 2 198 69
X70392.1 - 1 - 2 183 69

View Full Product Details