The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164171.1 | NP_001157643.1 | 20 - 21 | 2635 | 67 |
NM_010856.4 | NP_034986.1 | 20 - 21 | 2571 | 67 | |
GenBank mRNA | AK142232.1 | - | 20 - 21 | 2608 | 67 |
BC061145.1 | - | 20 - 21 | 2604 | 67 | |
BC090979.1 | - | 20 - 21 | 2604 | 67 | |
BC110700.1 | - | 20 - 21 | 2617 | 67 | |
BC138027.1 | - | 19 - 20 | 2510 | 67 | |
M76598.1 | - | 18 - 19 | 2433 | 67 | |
M76599.1 | - | 18 - 19 | 2433 | 67 | |
M76600.1 | - | 18 - 19 | 2433 | 67 | |
M76601.1 | - | 20 - 21 | 2535 | 67 |