The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038839.2 | NP_001033928.1 | 5 - 6 | 692 | 64 |
NM_001038845.2 | NP_001033934.1 | 5 - 6 | 692 | 64 | |
NM_001284402.1 | NP_001271331.1 | 5 - 6 | 520 | 64 | |
NM_011027.3 | NP_035157.2 | 5 - 6 | 692 | 64 | |
GenBank mRNA | AJ009823.1 | - | 5 - 6 | 540 | 64 |
AJ489296.1 | - | 5 - 6 | 529 | 64 | |
AJ489297.1 | - | 5 - 6 | 529 | 64 | |
AK144585.1 | - | 5 - 6 | 693 | 64 | |
AK145050.1 | - | 5 - 6 | 547 | 64 | |
AK145211.1 | - | 5 - 6 | 556 | 64 | |
BC128303.1 | - | 5 - 6 | 536 | 64 | |
BC128304.1 | - | 5 - 6 | 536 | 64 | |
BC141120.1 | - | 5 - 6 | 559 | 64 | |
FJ436444.1 | - | 5 - 6 | 520 | 64 |