Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkcg
Entrez Gene ID
18752
Gene Name
protein kinase C, gamma
Gene Aliases
PKCgamma, Pkcc, Prkcc
Chromosome Location
Chr.7: 3303532 - 3331099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291434.1 NP_001278363.1 11 - 12 1478 122
NM_011102.4 NP_035232.1 12 - 13 1631 122
GenBank mRNA AK144419.1 - 12 - 13 1633 122
BC111807.1 - 11 - 12 1365 122
L28035.1 - 12 - 13 1709 122
X67129.1 - 12 - 13 1440 122

View Full Product Details