The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011819.2 | NP_035949.2 | 1 - 2 | 303 | 72 |
GenBank mRNA | AF159571.1 | - | 1 - 2 | 774 | 72 |
AK144199.1 | - | 2 - 3 | 602 | 72 | |
AK159242.1 | - | 1 - 2 | 304 | 72 | |
AK160927.1 | - | 1 - 2 | 216 | 72 | |
BC067248.1 | - | 1 - 2 | 292 | 72 |