The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201470.1 | NP_001188399.1 | 6 - 7 | 889 | 74 |
NM_011864.3 | NP_035994.2 | 6 - 7 | 910 | 74 | |
GenBank mRNA | AF052453.1 | - | 6 - 7 | 891 | 74 |
AF085144.1 | - | 6 - 7 | 821 | 74 | |
AK128962.1 | - | 6 - 7 | 910 | 74 | |
AK137675.1 | - | 6 - 7 | 815 | 74 | |
BC090997.1 | - | 6 - 7 | 910 | 74 |